Order Entry
ContactUsLinkComponent
Square Laboratory Bottle
VITLAB® Square Laboratory Bottles, HDPE, Wide Mouth, with PP Screw Caps, BrandTech
Catalog #: 83009-056
Supplier:  Brandtech
Square Laboratory Bottle
VITLAB® Square Laboratory Bottles, HDPE, Wide Mouth, with PP Screw Caps, BrandTech
Catalog #: 83009-056
Supplier:  Brandtech

Specifications

  • Cap Size:
    GL50
  • Capacity:
    250 ml (8.5 oz.)
  • Dimensions:
    80 L x 80 W x 83 H mm
  • Cat. no.:
    83009-056
  • Supplier No.:
    V92689

Specifications

About this item

General-use, high-density polyethylene laboratory bottles come with polypropylene screw caps.

  • Feature a space-saving square shape
  • Wide mouth design provides excellent access to bottle contents

Bottles provide excellent chemical resistance to aliphatic and aromatic hydrocarbons, alcohols, aldehydes, alkalis, and acids.

Customers Who Bought This Also Bought