Order Entry
ContactUsLinkComponent
Polypropylene Graduated Pitcher
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Catalog #: 83008-752
Supplier:  Brandtech
Polypropylene Graduated Pitcher
VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Catalog #: 83008-752
Supplier:  Brandtech

Specifications

  • Capacity:
    2 L (68 oz.)
  • Graduation Interval:
    50 ml
  • Cat. no.:
    83008-752
  • Supplier No.:
    V443941

Specifications

About this item

Made of durable, translucent polypropylene.

  • Pitchers with molded graduations areavailable with or without overprint
  • Nesting pitchers have screened overprint
  • Blue printing provides high contrast with many liquids

Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.

Customers Who Bought This Also Bought