Inoculating Loops and Needles, Sterile, Greiner Bio-One
Catalog #: 82051-144
Supplier: Greiner Bio-One
Specifications
- Length:20 cm (77/8")
- Color:Blue
- Description:10 µL Inoculating Loops
- Cat. no.:82051-144
- Packaging:Individually Packed
- Supplier No.:731171
Specifications
About this item
Inoculating loops allow easier bacterial collection and inoculation.
- Color-coded
- Polystyrene construction
- Sterile
Inoculating needles are ideal for picking up single colonies.
Customers Who Bought This Also Bought