Order Entry
ContactUsLinkComponent
CELLSTAR Dish
CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Catalog #: 82050-916
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
CELLSTAR Dish
CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Catalog #: 82050-916
Supplier:  Greiner Bio-One
Product available on GSA Advantage®

Specifications

  • Ø:
    100 mm
  • Height:
    20 mm
  • Cat. no.:
    82050-916
  • Growth Area:
    58 cm²
  • Supplier No.:
    664160

Specifications

About this item

CELLSTAR® dishes are manufactured from high-quality virgin polystyrene and are treated for optimal cell attachment and proliferation.

  • High-quality virgin polystyrene construction
  • Nonpyrogenic
  • Free of RNase and DNase
  • Stackable
  • Uniform gas exchange and reduced condensation

Dishes offer distortion-free clarity. They are nonpyrogenic and RNase- and DNase-free. All dishes feature stacking rims for simple storage and ease in handling. Vented versions provide uniform gas exchange and reduced condensation.

Customers Who Bought This Also Bought