Order Entry
ContactUsLinkComponent
Cryo.s Tube
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Catalog #: 82050-212
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Cryo.s Tube
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Catalog #: 82050-212
Supplier:  Greiner Bio-One
Product available on GSA Advantage®

Specifications

  • Cap included:
    Yes
  • Capacity:
    2.00 ml
  • Description:
    Vials with writing area, external thread
  • Cat. no.:
    82050-212
  • Cap Color:
    Blue
  • Bottom Style:
    Round bottom, self-standing
  • Supplier No.:
    126279

Specifications

About this item

Designed for short- and long-term storage of samples and cells, these cryogenic storage vials are made from virgin polypropylene resin for purity, clarity, and consistency.

  • Nonpyrogenic and RNase- and DNase-free to <0.06 EU/ml

Cryo.s™ vials are available with internal or external thread. The internal thread format features a silicone gasket and is ideally suited for storage in the vapor phase of liquid nitrogen, at temperatures from –196 to 121 °C.

Delivery: All vials are supplied with caps. Each case includes cap inserts: 150 cap inserts come with cases of 500 and 100 come with cases of 100.
Customers Who Bought This Also Bought