VWR® Pipette Sterilization Boxes, Stainless Steel
Catalog #: 82027-608
Supplier: VWR International
Specifications
- Dimensions:5.1W×5.1D×41.9H cm (2×2×16½")
- Description:Square
- Cat. no.:82027-608
Specifications
About this item
Pipette sterilization boxes are designed to secure and protect pipettes during sterilization processes.
- Stainless steel construction
- High corrosion resistance
- Compatible with a variety of pipette types
Stainless steel construction ensures high corrosion resistance. Multiple box styles accommodate a variety of pipette types.
Customers Who Bought This Also Bought