Order Entry
ContactUsLinkComponent
VWR 6 inch PIPET BOX
VWR® Pipette Sterilization Boxes, Stainless Steel
Catalog #: 82027-606
Supplier:  VWR International
VWR 6 inch PIPET BOX
VWR® Pipette Sterilization Boxes, Stainless Steel
Catalog #: 82027-606
Supplier:  VWR International

Specifications

  • Dimensions:
    5.1W×5.1D×15.2H cm (2×2×6")
  • Description:
    Square (Pasteur)
  • Cat. no.:
    82027-606

Specifications

About this item

Pipette sterilization boxes are designed to secure and protect pipettes during sterilization processes.

  • Stainless steel construction
  • High corrosion resistance
  • Compatible with a variety of pipette types

Stainless steel construction ensures high corrosion resistance. Multiple box styles accommodate a variety of pipette types.

Customers Who Bought This Also Bought