VWR® Round/Tapered Micro Spatulas, Stainless Steel
Catalog #: 82027-520
Supplier: VWR International
Specifications
- Length:16.5 cm (6¹/₂")
- Cat. no.:82027-520
- Blade Length:—
Specifications
About this item
Ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
- Mirror finish
- Rustproof
- Non-sticking
Stainless steel spatulas feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
Customers Who Bought This Also Bought