VWR® Lab Scoop, Stainless Steel
Catalog #: 82027-498
Supplier: VWR International
Specifications
- Length:16.5 cm (61/2")
- Description:Scoop
- Cat. no.:82027-498
Specifications
About this item
The rounded tip of this scoop makes it easy to push into powders or break up solidified chemicals into smaller pieces, while its concave shape makes it ideal for picking them up.
- Sturdy stainless steel construction
- Handle and scoop sold separately
Lab scoops are sold in separate handle and scoop components to readily replace parts inexpensively.
Customers Who Bought This Also Bought