Order Entry
ContactUsLinkComponent
VWR SPATULA WOODEN HANDLE 6IN PK12 CS60
VWR® Spatulas with Wooden Handles
Catalog #: 82027-462
Supplier:  VWR International
VWR SPATULA WOODEN HANDLE 6IN PK12 CS60
VWR® Spatulas with Wooden Handles
Catalog #: 82027-462
Supplier:  VWR International

Specifications

  • Description:
    Standard blade spatulas
  • Cat. no.:
    82027-462
  • Blade Length:
    152 mm (6")
  • Handle Length:
    12.70 cm (5)"

Specifications

About this item

Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.

  • Durable
  • Stainless steel construction
  • Wooden handles

Multiple sizes allow for varied laboratory applications.

Customers Who Bought This Also Bought