VWR® Dumont Forceps
Catalog #: 82027-426
Supplier: VWR International
Specifications
- Length:114 mm (41/2")
- Cat. no.:82027-426
Specifications
About this item
Ideal for use with specimen screens for electron microscopes and electronic parts.
- Stainless steel fabrication
- Sharpened to a very fine point
Dumont forceps are ergonomically designed for comfortable use in a variety of applications.
Customers Who Bought This Also Bought