ContactUsLinkComponent
VWR* Antimagnetic Forceps
VWR® Antimagnetic Forceps
Catalog #: 82027-400
Supplier:  VWR International
VWR* Antimagnetic Forceps
VWR® Antimagnetic Forceps
Catalog #: 82027-400
Supplier:  VWR International
Specifications
  • Length:
    114 mm (41/2")
  • Cat. no.:
    82027-400
Specifications
About this item
Antimagnetic forceps are constructed from anti-magnetic stainless steel.
  • Good resistance to corrosion
  • Autoclavable
  • For general-purpose use
Forceps feature an overall length of 114 mm (4.5") and are available in a variety of unit sizes, from individual forceps to bulk packages.
Customers Who Bought This Also Bought