VWR® Dissecting Forceps, Medium Tip, Serrated
Catalog #: 82027-398
Supplier: VWR International
Specifications
- Length:114 mm (41/2")
- Cat. no.:82027-398
Specifications
About this item
VWR® Dissecting Forceps are stainless steel with straight, medium, serrated tips.
- Stainless steel fabrication
- Straight, medium-sized, serrated tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Customers Who Bought This Also Bought