VWR® Specimen Forceps
Catalog #: 82027-382
Supplier: VWR International
Specifications
- Length:305 mm (12")
Specifications
About this item
Specimen forceps are ergonomically designed for safely and efficiently removing specimens from deep jars or bottles.
- Stainless steel construction
- Straight tips
- Easy to use
Durable specimen forceps are built to withstand years of regular use.
Customers Who Bought This Also Bought