Order Entry
ContactUsLinkComponent
VWR* Small Latex Bulbs
VWR® Small Latex Bulbs
Catalog #: 82024-550
Supplier:  VWR International
VWR* Small Latex Bulbs
VWR® Small Latex Bulbs
Catalog #: 82024-550
Supplier:  VWR International

Specifications

  • Capacity:
    1 ml
  • Length:
    38 mm
  • Cat. no.:
    82024-550

Specifications

About this item

Small-volume, natural rubber latex bulbs are designed for use with Pasteur pipettes, small pipettes, and medicine droppers.

  • Small-volume design
  • Natural rubber latex construction
  • White finish

Supplied with white finish.

Customers Who Bought This Also Bought