ContactUsLinkComponent
VWR* Mechanical Timer
VWR® 60-Minute Mechanical Timer
Catalog #: 82023-840
Supplier:  VWR International
VWR* Mechanical Timer
VWR® 60-Minute Mechanical Timer
Catalog #: 82023-840
Supplier:  VWR International
Specifications
  • Cat. no.:
    82023-840
  • W×H:
    57×57 mm (2.2×2.2")
  • Accuracy:
    3 minutes
Specifications
About this item
Dial timer has a timing capacity of 60 minutes.
  • Counts down from 60 minutes
  • Audible alarm
  • Highly visible red arrow
Customers Who Bought This Also Bought