Orders LinkContactUsLinkComponent
VWR® Micro Slide Trays, Cardboard
VWR® Micro Slide Trays, Cardboard
20.8 x 34.1 x 9 cm (83/16 x 137/16 x 31/2"), 20
Catalog # 82020-913
Supplier:   VWR International
VWR® Micro Slide Trays, Cardboard
Catalog # 82020-913
Supplier:  VWR International
VWR® Micro Slide Trays, Cardboard
VWR® Micro Slide Trays, Cardboard
20.8 x 34.1 x 9 cm (83/16 x 137/16 x 31/2"), 20
Price
$34.13
New Customer Promotional Price
Log in for availability
Each Case of 72
Product Details & Documents
VWR® Micro Slide Trays accommodate 20 or 30 standard 25x75 mm (1x3") slides.
  • Sturdy cardboard construction
  • Recessed compartments protect specimens
  • Designed to allow easy slide insertion and removal
The horizontal compartments are recessed, protecting the specimens when the cover is closed. Compartments feature a thumb cut for easy slide removal.
Specifications
  • No. of places:
    20
  • L×W×H:
    20.8 x 34.1 x 9 cm (83/16 x 137/16 x 31/2")