VWR® Light-Duty Tissue Wipers
Catalog #: 82003-820
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- W×L:41/2×83/8"
- Description:1-ply light-duty tissue wipers in perforated box
- Packaging:280/box
Specifications
About this item
Virgin fiber tissue wipers are absorbent, soft, strong, and low-linting.
- Light-duty tissue wipers
- Designed for laboratory, pharmaceutical, and biomedical applications
- Constructed of virgin fiber papers
- Supplied in anti-static film windowed boxes
For use in laboratory, electronic, pharmaceutical, biomedical, optical, healthcare, computer assembly, graphic, and many other light manufacturing and assembly applications. Custom-size dispensers (available separately) hold wiper boxes securely, providing one-handed access to wipers.
Customers Who Bought This Also Bought