VWR® Slide Mailers for 5 Slides
Catalog #: 82003-431
Supplier: VWR International
Specifications
- Color:Assorted
- Description:End-Opening Mailer
- W×D×H:43 x 24 x 88 mm (111/16 x 15/16 x 315/32")
Specifications
About this item
Ideal for shipping, storing, or staining slides, these polypropylene containers hold five standard 25x75 mm (1x3") slides.
Inside wall features slotted channels to keep slides separated and make removal easier. Closes securely to keep slides safe.
Ordering information: Assorted colors include red, yellow, blue, purple, and orange.
Customers Who Bought This Also Bought