VWR® Microscope Slide Boxes for 100 Slides
Catalog #: 82003-414
Supplier: VWR International
Specifications
- Color:White
- Cat. no.:82003-414
Specifications
About this item
Each box features cork lining and an outer shell made of durable ABS plastic.
- Durable ABS plastic outer shell
- Stores up to 100 standard (1x3") slides
Secured with a rust-resistant nickel-plated clasp and hinge pin. Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. For storage of up to 100 standard 25x75mm (1x3") slides. Boxes stack securely on top of each other. Color-coded for easy identification. Dimensions: 22.2Lx17.1Wx3.3Dcm (83/4x63/4x15/16").
Customers Who Bought This Also Bought