Order Entry
ContactUsLinkComponent
Disposable Polyethylene Aprons
AlphaTec™ 56-230 Disposable Polyethylene Aprons, Ansell
Catalog #: 80093-298
Supplier:  Ansell Healthcare
AlphaTec™ 56-230 Disposable Polyethylene Aprons, Ansell
Catalog #: 80093-298
Supplier:  Ansell Healthcare

Specifications

  • Garment application:
    Food,
    Industrial
  • W×L:
    69×140 cm (27×55")
  • Color:
    Blue
  • Thickness:
    1.25 mils
  • Cat. no.:
    80093-298
  • Supplier No.:
    950145

Specifications

About this item

These disposable die-cut polyethylene aprons offer a combination of economical protection and flexible comfort for light-to-medium duty applications.

  • Convenient and adjustable fit
  • Inexpensive and hassle-free
  • Economical protection for light- or medium-duty applications

Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact

Use in the food processing, food service, and janitorial/sanitation industries. Uses include food handling and preparation and maintenance.

Polyethylene aprons traditionally offer excellent protection and resistance to chemicals, fats, oils and grease. Integral material ties provide convenience and an adjustable fit. Inexpensive enough to be discarded but strong enough to be reused if needed. The disposable polyethylene series is economical and conveniently packaged in a dispenser bag, providing a hassle-free experience.

Customers Who Bought This Also Bought