ContactUsLinkComponent
SYRINGE GLASS 2ML GLASS TIP
Poulten and Graf Fortuna™ Glass Syringes, Air-Tite Products
Catalog #: 80089-546
Supplier:  Air-Tite
Small Business Enterprise
SYRINGE GLASS 2ML GLASS TIP
Poulten and Graf Fortuna™ Glass Syringes, Air-Tite Products
Catalog #: 80089-546
Supplier:  Air-Tite
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    2 ml
  • Description:
    Glass Syringes
  • Cat. no.:
    80089-546
  • Packaging:
    1/box
  • For Use With:
    Laboratory and Veterinary Use Only
  • Type:
    Glass Luer Slip Tip
  • Supplier No.:
    GT2
Specifications
About this item
Fortuna Optima syringes are specialty syringes made from very fine borosilicate glass.
  • Borosilicate glass
  • Interchangeable pistons and barrels
  • Available with Luer Slip Tip or Luer Lock Tip
Fortuna Optima syringes are specialty syringes made from very fine borosilicate glass.
These syringes are manufactured in Germany from very fine borosilicate glass with interchangeable pistons and barrels. They feature amber graduations and have Luer lock and slip tips.
Certifications: Meets ISO Standards.
Ordering information: Individually boxed.
Customers Who Bought This Also Bought