Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog #: 80086-062
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:1200 µl
- Cat. no.:80086-062
- Sterile:+
- Supplier No.:TF-1200-C-R-S
Specifications
About this item
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought