Order Entry
ContactUsLinkComponent
GAS DISPERSION TUBE, FRITTED
Gas Dispersion Tubes, Fritted, Chemglass
Catalog #: 80073-612
Supplier:  Chemglass
Small Business Enterprise
GAS DISPERSION TUBE, FRITTED
Gas Dispersion Tubes, Fritted, Chemglass
Catalog #: 80073-612
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Porosity:
    Medium
  • Description:
    Gas Dispersion Tube, Medium Frit
  • Supplier No.:
    CG-203-02

Specifications

About this item

Tubes have a hollow fritted cylinder at the end of an 8 mm O.D. tube.

Cylinder dimensions: 120O.D.x20L mm (1/2x3/4"). Overall tube length: 254 mm (10").

Customers Who Bought This Also Bought