Order Entry
ContactUsLinkComponent
SPOON, MICRO
Spatulas, Spoon, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Catalog #: 80071-672
Supplier:  Chemglass
Small Business Enterprise
SPOON, MICRO
Spatulas, Spoon, Micro, #304 Stainless Steel, Non-Magnetic, Chemglass
Catalog #: 80071-672
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Length:
    16.5 cm (6.5")
  • Description:
    Stainless steel spatula, #304, non-magnetic, spoon style
  • Cat. no.:
    80071-672
  • Supplier No.:
    CG-1983-12

Specifications

About this item

Spoon-type spatula is made of non-magnetic stainless steel with an overall length of 6.5" (165 mm).

  • Spatula will pass through a 14/10 or 14/20 outer joint
  • The oval spoon end is ³/₄" (19 mm) long × ³/₁₆" (4.8 mm) wide
  • Equipped with a cushion grip vinyl handle for easy holding
Customers Who Bought This Also Bought