Freeze Dry Flasks, Chemglass
Catalog #: 80070-442
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:900 ml
- Description:Freeze Dry Flasks
- Cat. no.:80070-442
- For Use With:Labconco 7542900
- Supplier No.:CG-1611-07
Specifications
About this item
These flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers.
Wide mouth design provides easy access sample.
Ordering information: Flasks only, not suppled with rubber top, filter or O-ring.
Caution: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.
Customers Who Bought This Also Bought