Volumetric Flasks, Mixing Bulb, Class A, Chemglass
Catalog #: 80070-376
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:250 ml
- Cat. no.:80070-376
- ST Stopper No.:16
- Supplier No.:CG-1601-06
Specifications
About this item
Flasks feature a mixing bulb between the calibration line and stopper to aid in the mixing or dissolving of the contents and a solid glass pennyhead stopper
Conforms to Federal Specifications NNN-F-289 for Type 1, Class A unserialized flasks and ASTM E288.
The mixing bulb contains approximately one-tenth of the flask volume.
Customers Who Bought This Also Bought