Flasks, Erlenmeyer, Iodine, Stoppers, Chemglass
Catalog #: 80070-168
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:125 ml
- Description:Erlenmeyer Flask, Iodine, Stopper, Flask Only
- Cat. no.:80070-168
- Supplier No.:CG-1552-02
Specifications
About this item
Flasks feature funnel top with stoppers.
- A standard taper outer grind
- Accepts a 24/40 long neck stopper
- Used in ASTM D29 and ASTM D555
Available either as complete with stopper or flask only. Handle extends above the liquid seal.
Customers Who Bought This Also Bought