Rotary Evaporator Bump Traps, Fritted, Chemglass
Catalog #: 80068-126
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Bump Trap, Fritted, Rotary Evaporator
- Cat. no.:80068-126
- Bottom Inner [ST] Joint Size:24/40
- Supplier No.:CG-1323-21
Specifications
About this item
Rotary evaporator Bump Traps are designed with an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping.
The top outer [ST] joint on all traps is 24/40.
Customers Who Bought This Also Bought