Rotary Evaporator Adapters, Vial, Chemglass
Catalog #: 80068-042
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Glass Adapter, Top Outer [ST] Joint No. 24/40
- Cat. no.:80068-042
- Supplier No.:CG-1318-10
Specifications
About this item
Adapters permit the connection of standard laboratory dram vials directly to the vapor tube or bump trap of a rotary evaporator.
- Adapters have a 20-400 GPI thread finish at the bottom
- A fluoropolymer resin lined, polypropylene double-threaded connector attaches the vial to the adapter
Ordering information: The glass adapter and double-threaded connections must be ordered separately. Connector set includes one of each of the plastic connectors.
Customers Who Bought This Also Bought