Distilling Columns, Hemple, Chemglass
Catalog #: 80067-478
Supplier: Chemglass
Small Business Enterprise
Specifications
- Dimensions:10×130 mm
- Description:Distilling Column, Hemple
- Cat. no.:80067-478
- Joint Size:14/20
- Supplier No.:CG-1228-05
Specifications
About this item
Distillations feature a row of indentations.
- Plain distillation column
- Includes a row of indentations to support column packing material
Column length listed is for the straight portion above the row of indentations and below the joint. Column has a standard taper top outer and lower inner joint.
Customers Who Bought This Also Bought