Order Entry
ContactUsLinkComponent
REACTION TUBE, AIRFREE
Airfree® Schlenk Reaction Tubes, Single Neck, [ST] Inner Joint, 2 mm Stopcock, Chemglass
Catalog #: 80064-748
Supplier:  Chemglass
Small Business Enterprise
REACTION TUBE, AIRFREE
Airfree® Schlenk Reaction Tubes, Single Neck, [ST] Inner Joint, 2 mm Stopcock, Chemglass
Catalog #: 80064-748
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Capacity:
    250 ml
  • ST Joint No.:
    24/40 (inner joint)
  • Description:
    Airfree® Schlenk Reaction Tube with 2 mm Stopcock
  • Cat. no.:
    80064-748
  • Supplier No.:
    AF-0538-04

Specifications

About this item

Cylindrical shaped tube, with a standard taper inner joint, useful for simple reactions.

  • 2 mm high vacuum glass stopcock
  • Stainless steel constant pressure retaining device
Customers Who Bought This Also Bought