Order Entry
ContactUsLinkComponent
FLASK, 2-NECK, REACTION, AIRFREE
Airfree® Schlenk 2-Neck Reaction Flasks, Chemglass
Catalog #: 80064-696
Supplier:  Chemglass
Small Business Enterprise
FLASK, 2-NECK, REACTION, AIRFREE
Airfree® Schlenk 2-Neck Reaction Flasks, Chemglass
Catalog #: 80064-696
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Capacity:
    250 ml
  • ST Joint No.:
    24/40
  • Description:
    Airfree® Schlenk 2-Neck Reaction Flask, with 2 mm Glass Stopcock
  • Cat. no.:
    80064-696
  • Supplier No.:
    AF-0530-04

Specifications

About this item

Round bottom, two neck flasks have standard taper outer joints, both being the same size.

  • Equipped with either a 2 mm high vacuum glass stopcock or a 0 to 4 mm Chem-Cap® Airfree® inlet valve
  • Available in 14/20 or 24/40 joint sizes

Available with a choice of a 2 mm high vacuum glass stopcock with a stainless steel constant pressure retaining device, or a 0 to 4 mm Chem-Cap® Airfree® inlet valve. Side neck can be fitted with septum stopper for sampling or cannula transfer.

Customers Who Bought This Also Bought