Order Entry
ContactUsLinkComponent
AIRFREE* Reaction Flask
Airfree® Reaction Flasks, Kjeldahl Shaped, Chemglass
Catalog #: 80064-594
Supplier:  Chemglass
Small Business Enterprise
AIRFREE* Reaction Flask
Airfree® Reaction Flasks, Kjeldahl Shaped, Chemglass
Catalog #: 80064-594
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Capacity:
    100 ml
  • ST Joint No.:
    24/40
  • Cat. no.:
    80064-594
  • Supplier No.:
    AF-0520-05

Specifications

About this item

Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids

Lower portion of flask fits standard heating mantles. Comes with a 2mm glass stopcock, constant pressure retaining device, and an all fluoropolymer resin AIRFREE® valve.

Customers Who Bought This Also Bought