Glass Plates, Disc, Borosilicate Glass, Chemglass
Catalog #: 80061-706
Supplier: Chemglass
Small Business Enterprise
Specifications
- Ø:4"
- Description:Thick Borosilicate Disc
- Thickness:1/4"
- Cat. no.:80061-706
- Supplier No.:CG-1906-17
Specifications
About this item
Pyrex® or Duran® borosilicate glass disc.
- Edges on all sizes are ground and chamfered
Customers Who Bought This Also Bought