Order Entry
ContactUsLinkComponent
EXTRACTION APPARATUS, JUMBO
Accessories for Reaction Vessels, Chemglass
Catalog #: 80061-184
Supplier:  Chemglass
Small Business Enterprise
EXTRACTION APPARATUS, JUMBO
Accessories for Reaction Vessels, Chemglass
Catalog #: 80061-184
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Description:
    O-Ring, FEP-encased Silicone
  • Cat. no.:
    80061-184
  • For Use With:
    150 mm Flange
  • Supplier No.:
    CG-147-43

Specifications

About this item

Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring.

It also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease.

In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination.

Outer taper of flange is formed at a 45° angle to accept the CG-141 clamp.

Customers Who Bought This Also Bought