Inlet Thermometer Adapters, PTFE, Chemglass
Catalog #: 80060-308
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Inlet Thermometer Adapter, PTFE, for Joint Sizes 24/25 - 24/40
- Cat. no.:80060-308
- ST Inner Joint:24/40
- Supplier No.:CG-1048-01
Specifications
About this item
These PTFE adapters have an inner joint of the listed size.
- Made entirely of PTFE
The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7 mm.
Delivery: Supplied complete with one #108 Viton O-Ring.
Customers Who Bought This Also Bought