Nalgene® Replacement Coupling Inserts for Quick Filling Venting Closure; PP, Thermo Scientific
Catalog #: 73520-274
Supplier: Thermo Fisher Scientific
Specifications
- Tubing I.D.:6.4 mm (¼")
- Description:Straight
- Cat. no.:73520-274
- Supplier No.:2159-0001
Specifications
About this item
Nalgene Replacement coupling inserts support one handed connection and disconnection when applied to mating couple bodies
They promote fewer leakpoints, short assemblies, and faster installations.
Customers Who Bought This Also Bought