BioRobotix® PerkinElmer/Packard Tips, Thermo Scientific
Catalog #: 72830-514
Supplier: Thermo Scientific
Specifications
- Volume:20 µl
- Supplier No.:925-262
Specifications
About this item
Molecular BioProducts PerkinElmer/Packard pipette tips are designed to fit PerkinElmer/Packard MultiPROBE II HT and HT EX, PlateTrak, MiniTrak and Evolution P-3 workstations.
- Available in nonsterile or sterile formats
Each pipette tip is lot tested during production using a 15-point QC program on the actual workstation for which they are created. They have a low percentage Coefficient of Variance (%CV). The pipette tips, racks, and lids reduce static charges so tips pick up straight and eject perfectly each time.
They are also available with ART® filter technology and low retention design for critical samples, providing increased precision for the toughest multi-dispensing assays.
Customers Who Bought This Also Bought