BioRobotix® Pipette Tips for MultiMek™ Workstations, Molecular BioProducts
Catalog #: 72830-224
Supplier: Thermo Scientific
Specifications
- Volume:20 µl
- Tip Style:Standard
- Supplier No.:918-262
Specifications
About this item
Compatible with MultiMek™ style workstations/AP96.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as clear polypropylene only, in 20, 130 and 250 µl sizes. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
Ordering information: Tips are packaged 96 per tray, 10 trays per pack.
Customers Who Bought This Also Bought