ContactUsLinkComponent
55Mm Funnel
Buchner Funnel, Polypropylene, Ace Glass Incorporated
Catalog #: 71000-272
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
55Mm Funnel
Buchner Funnel, Polypropylene, Ace Glass Incorporated
Catalog #: 71000-272
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Funnel, Buchner, Polypropylene, Filter Paper Size 55 mm
  • Cat. no.:
    71000-272
  • Supplier No.:
    12557-07
Specifications
About this item
Lightweight polypropylene funnels
Bottom and top separate for easy cleaning.
Customers Who Bought This Also Bought