Order Entry
ContactUsLinkComponent
PADS, REPLACEMENT, 12/PKG, 72/CASE
SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Catalog #: 70690-182
PADS, REPLACEMENT, 12/PKG, 72/CASE
SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Catalog #: 70690-182

Specifications

  • Description:
    Replacement Pads
  • Cat. no.:
    70690-182
  • Supplier No.:
    362510000

Specifications

About this item

Mercury collector for capturing and collecting all spilled beads.

  • Container holds 125 mL of mercury
  • Use recommended disposal method
  • Replacement pads should be affixed with rubber cement
  • Dimensions are 10.5 O.D. x 4.8H cm (4⅛ x 2")

To collect spilled mercury quickly, unscrew the lid with the foam pad, and press pad firmly onto the spill, pressure forces the mercury into the pad. When the lid is screwed back onto the jar, the pad is compressed against the jar’s freestanding perforated plate, releasing the mercury into the jar bottom.

Customers Who Bought This Also Bought