SP Bel-Art Wash Bottle, 2-Color, Safety-Labeled, Wide-Mouth, LDPE, Bel-Art Products, a part of SP
Catalog #: 70690-074
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:500 ml (16.9 oz.)
- Description:Deionized Water
- Cat. no.:70690-074
- Cap Color:Blue
- Supplier No.:116460631
Specifications
About this item
Bel-Art’s Safety Labeled wash bottles for dispensing your most common solvents.
- Polypropylene closure with leakproof design
- Easy to read
- LDPE solvent-resistant
Printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making the workplace a safer place.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Ordering information: Assortment includes one each of Acetone, Ethanol, Isopropanol, Methanol, Toluene, and Water. Use only with solvents compatible with LDPE.
Customers Who Bought This Also Bought