ContactUsLinkComponent
AQUASONIC 100 TRANS GEL 5L CS4
AQUASONIC® 100 Ultrasound Transmission Gel, Parker Lab Inc.
Catalog #: 68200-712
Supplier:  Parker Lab
AQUASONIC® 100 Ultrasound Transmission Gel, Parker Lab Inc.
Catalog #: 68200-712
Supplier:  Parker Lab
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Aquasonic 100 gel
  • Cat. no.:
    68200-712
  • Packaging:
    5 liter SONICPAC® with refillable dispenser, 1 per box, 4 per case
  • Supplier No.:
    01-50
Specifications
About this item
Widely used gel for diagnostic and therapeutic medical ultrasound which is completely aqueous, will not stain clothing or damage transducers.
  • Acoustically correct for the broad range of frequencies used
  • Unique "can't be copied" formula is bacteriostatic, non-sensitizing and non-irritating
  • Applicable for all procedures where a viscous gel is required
  • No formaldehyde
  • Not a spermicide
Customers Who Bought This Also Bought