Order Entry
ContactUsLinkComponent
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Catalog #: 66115-154
Supplier:  GFS Chemicals
Small Business Enterprise
CAS Number:  
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Catalog #: 66115-154
Supplier:  GFS Chemicals
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 L bottle
  • Cat. no.:
    66115-154
  • For Use With:
    Non-Ratio Meters
  • Supplier No.:
    85092

Specifications

About this item

EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers

EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers

One year shelf life.

Ready to use at their stated value without mixing or diluting. Composed of submicron solid polymer beads suspended in ultra-pure water. Guaranteed accuracy within 1%. Safe, non-toxic, non-carcinogenic alternative to Formazin. Available for non-ratio and ratio meters.

Certifications: Approved by EPA, Standard Methods, ASTM, ASBC, and EBC. Traceable to NIST 1690, 1691.
Customers Who Bought This Also Bought