PYREX® Watch Glasses, Corning
Catalog #: 66110-021
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Ø:65 mm (29/16")
- Description:Watch glass
- Cat. no.:66110-021
- Supplier No.:9985-65
Specifications
About this item
PYREX® borosilicate glass.
- High chemical durability
- Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Customers Who Bought This Also Bought