Order Entry
ContactUsLinkComponent
NALGENE* Unwire* Polyketone Half-Rack
Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Catalog #: 66023-903
NALGENE* Unwire* Polyketone Half-Rack
Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Catalog #: 66023-903

Specifications

  • Color:
    White
  • Cat. no.:
    66023-903
  • L×W×H:
    12.8 x 10.3 x 8.3 cm (5 x 4 x 31/4")
  • For Tube O.D.:
    20 mm
  • Supplier No.:
    5972-0020

Specifications

About this item

These racks are ideal for applications using 36 tubes or less

Racks for 13mm and 16mm tubes hold 36 tubes; racks for 20mm and 25mm tubes hold 20 tubes; racks for 30mm tubes hold 9 tubes.

These racks are made of polyketone.

They are molded of ResMer® resin, which has excellent chemical resistance and can be immersed in a water bath. Racks feature excellent thermal properties which allow both autoclaving and freezing. Letters and numbers on the racks aid in tube identification.

Customers Who Bought This Also Bought