Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
Catalog #: 62662-048
Supplier: Thermo Fisher Scientific
Specifications
- Cat. no.:62662-048
- L×W×H:54.3 x 43.5 x 13 cm (213/8 x 171/8 x 51/8")
- Supplier No.:6900-0020
Specifications
About this item
Useful for drying and sterilizing instruments and labware, and for collecting and washing soiled lab equipment
Rolled edges for extra stiffness. Handles for easy carrying. Tapered for nesting. Autoclavable.
Customers Who Bought This Also Bought