Accessories for Microtiter® 96-Well Plates, Thermo Scientific
Catalog #: 62402-969
Supplier: Thermo Fisher Scientific
Specifications
- Description:Removawell Holder
- Cat. no.:62402-969
- Supplier No.:6604
Specifications
About this item
These accessories are for use with all Thermo Scientific Immulon®, Microfluor®, and Microlite™ Microtiter plates and Removawell* assemblies.
Rigid styrene lids are sterile. Flexible vinyl lids 62403-072 are for use with Removawell assemblies. Clear precut acetate, Mylar®, and PVC plate sealers are for storage and incubation procedures.
The Removawell holder 62402-969 holds eight 12-well or twelve 8-well Removawell strips in a 96-well format.
Customers Who Bought This Also Bought