Dounce Tissue Grinders, Wheaton
Catalog #: 62400-595
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Specifications
- Capacity:1 ml
- Cat. no.:62400-595
- Chamber Dimensions:11O.D. x 48L mm
- Supplier No.:357538
Specifications
About this item
Designed for fine particle size reduction without damage to cell nuclei
Made of low extractable borosilicate glass. Supplied with 2 pestles. The large clearance pestle is used for initial sample reduction; the narrow clearance pestle forms the final homogenate. Mortar has large reservoir and pouring lip.
For small cells such as E. coli, proteins, serum, and malignant tissues.
Loose pestle clearance: 0.0889 - 0.127 mm (0.0035 - 0.0050")
Tight pestle clearance: 0.0254 - 0.0762 mm (0.0010 - 0.0030")
Customers Who Bought This Also Bought